Pentyl-4-hydroxybenzoate ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Pentylparaben, 4-Hydroxybenzoic acid Amyl Ester, 4-Hydroxybenzoic acid pentyl ester, Pentyl parahydroxybenzoate, Pentyl 4-Hydroxybenzoate, 4-Hydroxybenzoic acid Pentyl Ester, Amyl 4-Hydroxybenzoate, Amylparaben, n-Pentyl 4-hydroxybenzoate

A151168-1G A151168-25G A151168-5G
ALA151168-1GEA 9.41 USD
ALA151168-1G ALA151168-25G ALA151168-5G
Pentyl-4-hydroxybenzoate ≥98%
Pentyl 4-hydroxybenzoate
Formula: C₁₂H₁₆O₃
Boiling Pt: 197-200°C
Melting Pt: 36-40°C
Storage Temperature: Refrigerator
MDL Number: MFCD00016479
CAS Number: 6521-29-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36f9a6b61a6dcb10e9877661cc834006c0b06643&vid=36f9a6b61a6dcb10e9877661cc834006c0b06643&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR