2-Amyl-5-methylphenol ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 5-Methyl-2-pentylphenol, Amylmetacresol

A151165-5G A151165-1G A151165-25G
ALA151165-5GEA 310.56 USD
ALA151165-5G ALA151165-1G ALA151165-25G
2-Amyl-5-methylphenol ≥98% (by GC)
2-Amyl-5-methylphenol
Formula: C₁₂H₁₈O
MW: 178.28 g/mol
Boiling Pt: 139°C
Flash Pt: 125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00127710
CAS Number: 1300-94-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a105063e48179a3d186e8d38590d9c9f7ba75fc3&vid=a105063e48179a3d186e8d38590d9c9f7ba75fc3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR