4-Aminoantipyrine hydrochloride ≥98% (by HPLC, titration analysis) for biochemical research
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
4-AA.HCl, ['4-AA·HCl'], 4-Aminoantipyrine Hydrochloride, 4-Amino-1,5-dimethyl-2-phenyl-1H-pyrazol-3(2H)-one hydrochloride
A151163-1G
A151163-5G
ALA151163-1GEA
11.31
USD
ALA151163-1G
ALA151163-5G
4-Aminoantipyrine hydrochloride ≥98% (by HPLC, titration analysis) for biochemical research
4-Aminoantipyrine hydrochloride
Formula:
C₁₁H₁₃N₃O MW: 239.70 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00042012 CAS Number: 22198-72-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...