(-)-ɑ-Phellandrene ≥65% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Product Introduction:(R)-(-)-α-Phellandrene is a monoterpene derivative.Product Application:(R)-(-)-α-Phellandrene undergoes [4+2] cycloaddition with 2-naphthyl acetylenecarboxylate to form a chiral diene, which can act as a ligand for the rhodium-catalyzed asymmetric addition reactions. It may be used to synthesize (1S,5R)-5-(1-methylethyl)-2-methylidene-3-cyclohexen-1-yl hydroperoxide, which on reduction forms trans-yabunikkeol.
Formula:
C₁₀H₁₆ Boiling Pt: 175°C Density: 0.844 g/cm³ Flash Pt: 49°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00062988 CAS Number: 4221-98-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...