3-Amino-2-naphthoic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A151154-1G A151154-5G
ALA151154-1GEA 39.81 USD
ALA151154-1G ALA151154-5G
3-Amino-2-naphthoic acid ≥97%
3-Amino-2-naphthoic acid

Application:3-Amino-2-naphthoic acid is a dyes analytical reagent which is used as an intermediate for organic synthesis.

Formula: C₁₁H₉NO₂
Melting Pt: 212°C
MDL Number: MFCD00004115
CAS Number: 5959-52-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1f9a0c0aa29dbe1b745c7b7f07acec9ea848680&vid=f1f9a0c0aa29dbe1b745c7b7f07acec9ea848680&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR