2-Acetamido-6-hydroxypurine ≥95% (by HPLC, titration analysis)

Supplier: ALADDIN SCIENTIFIC

Synonyms: N-2-Acetylguanine, N-Acetylguanine

A151132-1G A151132-25G A151132-5G
ALA151132-1GEA 160.46 USD
ALA151132-1G ALA151132-25G ALA151132-5G
2-Acetamido-6-hydroxypurine ≥95% (by HPLC, titration analysis)
2-Acetamido-6-hydroxypurine
Formula: C₇H₇N₅O₂
MW: 193.17 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00078201
CAS Number: 19962-37-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01334000a03bdc260e52a72e14434976209c9534&vid=01334000a03bdc260e52a72e14434976209c9534&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR