3-Aminobenzyl alcohol ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151125-100G A151125-25G A151125-5G
ALA151125-100GEA 592.71 USD
ALA151125-100G ALA151125-25G ALA151125-5G
3-Aminobenzyl alcohol ≥98% (by GC)
3-Aminobenzyl alcohol

Product Application:3-Aminobenzyl alcohol, is used as a reagent in the synthesis of quinone analogs as dynamin GTPase inhibitors. Also used as a reagent in the synthesis of pyrrolo[2,1-f][1,2,4]triazines as novel hedgehog signaling pathway inhibitors.

Formula: C₇H₉NO
Melting Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00007817
CAS Number: 1877-77-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e2c9e0093c39b1a9e377cfa86bd6d4faf4c7382&vid=5e2c9e0093c39b1a9e377cfa86bd6d4faf4c7382&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR