2-(4-Aminophenyl)ethanol ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151120-100G A151120-25G A151120-5G
ALA151120-100GEA 227.91 USD
ALA151120-100G ALA151120-25G ALA151120-5G
2-(4-Aminophenyl)ethanol ≥98% (by GC)
2-(4-Aminophenyl)ethanol

4-Aminophenethyl alcohol has been used: . as nonsymmetric monomer in the preparation of ordered [head-to-head (H-H) or tail-to-tail (T-T)] poly(amide-ester) . in the synthesis of 4-aminostyrene . in functionalization of graphene nanoplatelets

Formula: C₈H₁₁NO
Boiling Pt: 255°C
Melting Pt: 107-110°C
Storage Temperature: Ambient
MDL Number: MFCD00007922
CAS Number: 104-10-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7feb92f1a261973fff758a7c4fd4b0a53248db4c&vid=7feb92f1a261973fff758a7c4fd4b0a53248db4c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR