ɑ,ɑ'-Bis(4-methylbenzyloxy)-p-xylene ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: p-Xylylene Glycol Bis(4-methylbenzyl) Ether, α,α'-Bis(4-methylbenzyloxy)-p-xylene

A151119-1G A151119-250MG
ALA151119-1GEA 197.51 USD
ALA151119-1G ALA151119250MG
ɑ,ɑ'-Bis(4-methylbenzyloxy)-p-xylene ≥98%
alpha,alpha'-Bis(4-methylbenzyloxy)-p-xylene
Formula: C₂₄H₂₆O₂
MW: 346.47 g/mol
Melting Pt: 100 °C
MDL Number: MFCD00143577
CAS Number: 136861-46-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb372c4b2f89f1524655b1dfbbaf9cd738458f17&vid=fb372c4b2f89f1524655b1dfbbaf9cd738458f17&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR