Magnesium 8-anilinonaphthalene-1-sulfonate hydrate ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 8-Anilino-1-naphthalenesulphonic acid magnesium(II) salt, Magnesium(II) 8-Anilino-1-naphthalenesulfonate, 8-Anilino-1-naphthalenesulfonic acid magnesium(II) salt, Magnesium 8-(phenylamino)naphthalene-1-sulfonate, ANS-Mg, 1,8-ANS magnesium salt, N-Phenyl peri acid magnesium salt, 8-Anilino-1-naphthalenesulfonic acid Magnesium(II) Salt Hydrate, ANSA magnesium salt, Magnesium(II) 8-Anilino-1-naphthalenesulfonate Hydrate, Magnesium(II) 8-Anilino-1-naphthalenesulphonate Hydrate, Magnesium 8-(phenylamino)naphthalene-1-sulphonate, Magnesium(II) 8-Anilino-1-naphthalenesulphonate, 8-Anilino-1-naphthalenesulphonic acid Magnesium(II) Salt Hydrate, 8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate, 8-Anilino-1-naphthalenesulphonic acid hemimagnesium salt hydrate

A151110-1G A151110-5G
ALA151110-1GEA 43.61 USD
ALA151110-1G ALA151110-5G
Magnesium 8-anilinonaphthalene-1-sulfonate hydrate ≥98% (by HPLC)
Magnesium 8-anilinonaphthalene-1-sulphonate hydrate

ANSA (8-Anilino-1-naphthalenesulfonic acid) is a hydrophobic polarity sensitive fluorescent dye useful as a site probe to detect conformational changes in cell and micelle membranes and molecules such as proteins. ANSA is also used as a fluorescent probe for the estimation of the critical micelle concentration (CMC) of surfactants.

Formula: C₃₂H₂₄MgN₂O₆S₂·xH₂O
MW: 620.98 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00065353
CAS Number: 18108-68-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1488faff5bfedabc262064d8de6597d6620c1762&vid=1488faff5bfedabc262064d8de6597d6620c1762&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR