Allyl-4-hydroxybenzoate 98

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Hydroxybenzoic Acid Allyl Ester, Allylparaben, Allyl 4-hydroxybenzoate

A151097-100G A151097-25G A151097-5G
ALA151097-100GEA 484.41 USD
ALA151097-100G ALA151097-25G ALA151097-5G
Allyl-4-hydroxybenzoate 98
Allyl-4-hydroxybenzoate
Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Melting Pt: 103 °C
MDL Number: MFCD08276268
CAS Number: 18982-18-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a1549c7917f529f6b136f6ae594b8ae59e95c57&vid=6a1549c7917f529f6b136f6ae594b8ae59e95c57&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR