Allyl-p-tolyl ether ≥95%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 4-Allyloxytoluene, Allyl p-Tolyl Ether, Allylp-tolylether

A151094-1G A151094-25G A151094-5G
ALA151094-1GEA 22.71 USD
ALA151094-1G ALA151094-25G ALA151094-5G
Allyl-p-tolyl ether ≥95%
Allyl-p-tolyl ether
Formula: C₁₀H₁₂O
MW: 148.21 g/mol
Flash Pt: 80 °C
MDL Number: MFCD00026098
CAS Number: 23431-48-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c51487729ba2ea0abaae6c44b4961cf1ec0a08c&vid=2c51487729ba2ea0abaae6c44b4961cf1ec0a08c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR