5-Acetylthiophene-2-carboxylic acid ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A151088-100G
A151088-10G
A151088-1G
A151088-25G
A151088-5G
ALA151088-100GEA
1895.16
USD
ALA151088-100G
ALA151088-10G
ALA151088-1G
ALA151088-25G
ALA151088-5G
5-Acetylthiophene-2-carboxylic acid ≥97%
5-Acetylthiophene-2-carboxylic acid
Formula:
C₇H₆O₃S Melting Pt: 208-215°C |
MDL Number:
MFCD00055512 CAS Number: 4066-41-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...