1-Acetylisatin ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 1-Acetylindoline-2,3-dione

A151084-1G A151084-25G A151084-5G
ALA151084-1GEA 20.81 USD
ALA151084-1G ALA151084-25G ALA151084-5G
1-Acetylisatin ≥97%
1-Acetylisatin
Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
Melting Pt: 143 °C
MDL Number: MFCD00158542
CAS Number: 574-17-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6b7cff7ed76133ee3a365fa2b05edbf9fd2b979&vid=b6b7cff7ed76133ee3a365fa2b05edbf9fd2b979&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR