3-Amino-4-methylbenzyl alcohol ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A151061-1G
A151061-250MG
A151061-25G
A151061-5G
ALA151061-1GEA
41.71
USD
ALA151061-1G
ALA151061250MG
ALA151061-25G
ALA151061-5G
3-Amino-4-methylbenzyl alcohol ≥98%
3-Amino-4-methylbenzyl alcohol
Formula:
C₈H₁₁NO Melting Pt: 107.0 to 111.0 °C |
MDL Number:
MFCD00075051 CAS Number: 81863-45-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...