4-Aminocyclohexaneethanol (cis- and trans- mixture) 95
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
1-Amino-4-(2-hydroxyethyl)cyclohexane, 4-Aminocyclohexaneethanol, 2-(4-Aminocyclohexyl)ethanol
A151060-1G
A151060-250MG
A151060-5G
ALA151060-1GEA
119.61
USD
ALA151060-1G
ALA151060250MG
ALA151060-5G
4-Aminocyclohexaneethanol (cis- and trans- mixture) 95
4-Aminocyclohexaneethanol (cis- and trans- mixture)
Formula:
C₈H₁₇NO MW: 143.23 g/mol Melting Pt: 85°C Storage Temperature: Ambient |
MDL Number:
MFCD15072144 CAS Number: 857831-26-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...