2-(2-Aminoethyl)-1,3-dioxolane ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A151047-5G A151047-1G A151047-250MG
ALA151047-5GEA 272.56 USD
ALA151047-5G ALA151047-1G ALA151047250MG
2-(2-Aminoethyl)-1,3-dioxolane ≥98%
2-(2-Aminoethyl)-1,3-dioxolane
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 75°C
Storage Temperature: Refrigerator
MDL Number: MFCD00142756
CAS Number: 5754-35-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=645f756e7ff96fddcba17632f1186f5bc50912b6&vid=645f756e7ff96fddcba17632f1186f5bc50912b6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR