Amyl-4-(4-Ethoxyphenoxycarbonyl)phenyl carbonate ≥95%

Supplier: ALADDIN SCIENTIFIC

Synonyms: Amyl 4-(4-Ethoxyphenoxycarbonyl)phenyl Carbonate, 4-(4-Ethoxyphenoxycarbonyl)phenyl Pentyl Carbonate

A151042-100MG A151042-250MG
ALA151042100MGEA 33.16 USD
ALA151042100MG ALA151042250MG
Amyl-4-(4-Ethoxyphenoxycarbonyl)phenyl carbonate ≥95%
Amyl-4-(4-Ethoxyphenoxycarbonyl)phenyl carbonate
Formula: C₂₁H₂₄O₆
MW: 372.42 g/mol
MDL Number: MFCD00059434
CAS Number: 33926-46-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e4e4bd798b50e5c97cdbaa3d92a47d68b075b53&vid=5e4e4bd798b50e5c97cdbaa3d92a47d68b075b53&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR