4-Amino-L-phenylalanine monohydrate ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Amino-L-phenylalanine Monohydrate, H-Phe(4-NH|2|)-OH Monohydrate, H-Phe(4-NH2)-OH Monohydrate

A151041-1G A151041-250MG A151041-5G
ALA151041-1GEA 131.01 USD
ALA151041-1G ALA151041250MG ALA151041-5G
4-Amino-L-phenylalanine monohydrate ≥98%
4-Amino-L-phenylalanine monohydrate
Formula: C₉H₁₂N₂O₂·H₂O
MW: 180.21 g/mol
Melting Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00150889
CAS Number: 304671-92-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9aef9d05aead226876acfcf4bacea1a6dc8838&vid=3f9aef9d05aead226876acfcf4bacea1a6dc8838&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR