Aminoguanidine hydrochloride ≥98% (by titration)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Aminoguanidine HCl, Aminoguanidine Hydrochloride, Guanylhydrazine hydrochloride

A151036-100G A151036-25G
ALA151036-100GEA 82.56 USD
ALA151036-100G ALA151036-25G
Aminoguanidine hydrochloride ≥98% (by titration)
Aminoguanidine hydrochloride

Product Introduction:

Aminoguanidine hydrochloride has been reported in a study as inhibitor of animal nitric oxide (NO)-synthase. Crystal structure of aminoguanidine hydrochloride has been investigated by Fourier and least squares method. Its crystals were monoclinic and the guanidine part of the aminoguanidinium ion is planar.


Product Usage

Aminoguanidine hydrochloride may be employed as nitric oxide synthase (NOS) inhibitor to investigate its effect on the reduction of alveolar bone loss in ligature induced periodontitis in rats. It may be used in the synthesis of 5-guanylhydrazone derivatives, having antibacterial activity against antibacterial activity against both Escherichia coli and Staphylococcus aureus.

Formula: CH₇ClN₄
Melting Pt: 162-166 °C
Storage Temperature: Ambient
MDL Number: MFCD00039074
CAS Number: 1937-19-5
EINECS: 217-707-7
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0f618b9efee8d6e791985956113f2df1182bcca&vid=c0f618b9efee8d6e791985956113f2df1182bcca&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR