a-Tosylbenzyl isocyanide ≥98% (by HPLC, total nitrogen)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: (1-Phenyl-1-tosyl)methyl Isocyanide, alpha-(p-Toluenesulfonyl)benzyl Isocyanide, α-Tosylbenzyl Isocyanide, α-(p-Toluenesulphonyl)benzyl Isocyanide, alpha-(p-Toluenesulphonyl)benzyl Isocyanide, alpha-Tosylbenzyl Isocyanide, α-(p-Toluenesulfonyl)benzyl Isocyanide

A151035-1G A151035-250MG A151035-5G
ALA151035-1GEA 524.31 USD
ALA151035-1G ALA151035250MG ALA151035-5G
a-Tosylbenzyl isocyanide ≥98% (by HPLC, total nitrogen)
a-Tosylbenzyl isocyanide
Formula: C₁₅H₁₃NO₂S
MW: 271.3342 g/mol
Melting Pt: 106 °C
MDL Number: MFCD04114776
CAS Number: 36635-66-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df881f30edeb357c347c0e3ebdfe64b85491be1a&vid=df881f30edeb357c347c0e3ebdfe64b85491be1a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR