ɑ,ɑ'-Dihydroxy-1,3-diisopropylbenzene 98 GC_ASSAY_METHOD

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 1,3-Bis(α-hydroxyisopropyl)benzene, α,α'-Dihydroxy-1,3-diisopropylbenzene, α,α,α',α'-Tetramethyl-1,3-benzenedimethanol, ɑ,ɑ,ɑ',ɑ'-Tetramethyl-1,3-benzenedimethanol, 1,3-Bis(ɑ-hydroxyisopropyl)benzene, 2,2'-(m-Phenylene)di-2-propanol

A151015-5G A151015-100G A151015-25G
ALA151015-5GEA 11.31 USD
ALA151015-5G ALA151015-100G ALA151015-25G
ɑ,ɑ'-Dihydroxy-1,3-diisopropylbenzene 98 GC_ASSAY_METHOD
alpha,alpha'-Dihydroxy-1,3-diisopropylbenzene
Formula: C₁₂H₁₈O₂
MW: 194.27 g/mol
Melting Pt: 138-142°C
MDL Number: MFCD00059139
CAS Number: 1999-85-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4cffa80590396671c7da47521f7892d54c68b89&vid=c4cffa80590396671c7da47521f7892d54c68b89&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR