9-Aminoacridine ≥97% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 9-AA

A151005-1G A151005-250MG A151005-5G
ALA151005-1GEA 48.36 USD
ALA151005-1G ALA151005250MG ALA151005-5G
9-Aminoacridine ≥97% (by HPLC)
9-Aminoacridine

Product Application:A MALDI matrix exhibiting low background and good sensitivity in the analysis of low molecular weight anions, such as metabolites, in negative mode

Formula: C₁₃H₁₀N₂
MW: 194.24 g/mol
Melting Pt: 238 °C
MDL Number: MFCD00037839
CAS Number: 90-45-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3909a613ae7c4d2b995572bcc9ea0bada50b846d&vid=3909a613ae7c4d2b995572bcc9ea0bada50b846d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR