5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide

A151003-25G A151003-5G
ALA151003-25GEA 196.56 USD
ALA151003-25G ALA151003-5G
5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide ≥97%
5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide
Formula: C₁₆H₂₀I₃N₃O₇
MW: 747.06 g/mol
Melting Pt: 275°C
MDL Number: MFCD08063354
CAS Number: 31127-80-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c7200895364c10f3520b020c475dbd5cbb204d9&vid=5c7200895364c10f3520b020c475dbd5cbb204d9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR