Angelicin ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
Furo[2,3-h]benzopyran-2-one, 2H-Furo[2,3-h]chromen-2-one, Isopsoralen, Bakuchicin, NSC 404563
A141501-10MG
A141501-25MG
ALA141501-10MGEA
22.71
USD
ALA141501-10MG
ALA141501-25MG
Angelicin ≥98%
Angelicin
An angular furocoumarin, may be used as a photoactivatable molecule that reacts with DNA and unsaturated fatty acids to form monoadducts. Angelicin may be studied as a potential phototherapeutic and to understand its mechanisms of action and effects on RNA and DNA synthesis.
Formula:
C₁₁H₆O₃ MW: 186.17 g/mol Boiling Pt: 104 °C Melting Pt: 140 °C |
MDL Number:
MFCD00064930 CAS Number: 523-50-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...