Astaxanthine cis- and trans-mixture ≥95% (by HPLC)

Supplier: ALADDIN SCIENTIFIC

Synonyms: Carophyll Pink, (3S,3'S)-Astaxanthin, Natupink, (6S,6'S)-3,3'-((1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-Tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaene-1,18-diyl)bis(6-hydroxy-2,4,4-trimethylcyclohex-2-enone), BioAstin

A141428-250MG A141428-50MG
ALA141428250MGEA 327.66 USD
ALA141428250MG ALA141428-50MG
Astaxanthine cis- and trans-mixture ≥95% (by HPLC)
Astaxanthine

Astaxanthin is a potent carotenoid antioxidant found in marine algae, red yeast and many other plant and animal sources. Animal studies indicate that it reduces blood glucose and ameliorates several parameters of the diabetic metabolic syndrome. It improves blood flow and vascular tone in models of hypertension. In vitro studies indicate that it upregulates connexin 43 and thus, may be chemopreventive against cancer.

Formula: C₄₀H₅₂O₄
MW: 596.852 g/mol
Melting Pt: 215-216°C
Storage Temperature: Ambient
MDL Number: MFCD00672621
CAS Number: 472-61-7
EINECS: 207-451-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=014981148c3810a194cc80ad4031cf429f4bcaf9&vid=014981148c3810a194cc80ad4031cf429f4bcaf9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR