4'-Isothiocyanatoacetophenone ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 4-Acetylphenyl isothiocyanate

A140590-1G A140590-5G
ALA140590-1GEA 98.71 USD
ALA140590-1G ALA140590-5G
4'-Isothiocyanatoacetophenone ≥98%
4'-Isothiocyanatoacetophenone
Formula: C₉H₇NOS
MW: 177.23 g/mol
Boiling Pt: 112°C
Melting Pt: 77-81°C
MDL Number: MFCD00041100
CAS Number: 2131-57-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29de3ac5fc1268d4fd26606138e661618e9f4c76&vid=29de3ac5fc1268d4fd26606138e661618e9f4c76&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR