Aluminium oxide ≥99.9% (metals basis)
Supplier: ALADDIN SCIENTIFIC
A140296-100G
A140296-500G
ALA140296-100GEA
9.41
USD
ALA140296-100G
ALA140296-500G
Aluminium oxide ≥99.9% (metals basis)
Aluminium oxide
Formula:
Al₂O₃ Boiling Pt: 2977°C Melting Pt: 2000°C Density: 1.06 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...