2-Amino-4-(4-bromophenyl)thiazole ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A140256-25G A140256-5G
ALA140256-25GEA 153.81 USD
ALA140256-25G ALA140256-5G
2-Amino-4-(4-bromophenyl)thiazole ≥97%
2-Amino-4-(4-bromophenyl)thiazole
Formula: C₉H₇BrN₂S
Melting Pt: 183-187°C
MDL Number: MFCD00051953
CAS Number: 2103-94-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a055bf6c47577d099b23ecb7f0c0d09461922ff&vid=0a055bf6c47577d099b23ecb7f0c0d09461922ff&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR