2-Amino-4-methylthiazole-5-carboxylic acid ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A140221-1G A140221-5G
ALA140221-1GEA 45.51 USD
ALA140221-1G ALA140221-5G
2-Amino-4-methylthiazole-5-carboxylic acid ≥95%
2-Amino-4-methylthiazole-5-carboxylic acid
Formula: C₅H₆N₂O₂S
Melting Pt: 166-170°C
Storage Temperature: Refrigerator
MDL Number: MFCD01853938
CAS Number: 67899-00-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4a787b4d80cd29ce3b0839c5ed6b6d033636c0c&vid=c4a787b4d80cd29ce3b0839c5ed6b6d033636c0c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR