5-Amino-4,6-dichloro-2-(propylthio)pyrimidine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4,6-Dichloro-2-(propylthio)pyrimidin-5-amine

A140069-100G A140069-1G A140069-250MG A140069-25G
ALA140069-100GEA 2676.06 USD
ALA140069-100G ALA140069-1G ALA140069250MG ALA140069-25G
5-Amino-4,6-dichloro-2-(propylthio)pyrimidine ≥98%
5-Amino-4,6-dichloro-2-(propylthio)pyrimidine
Formula: C₇H₉Cl₂N₃S
MDL Number: MFCD16875503
CAS Number: 145783-15-9
EINECS: 808-051-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a83502ffa27f2d64061f5a187723e80902d1f13&vid=2a83502ffa27f2d64061f5a187723e80902d1f13&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR