5-Amino-2-methylbenzenesulfonamide ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A139194-100G A139194-1G A139194-25G A139194-5G
ALA139194-100GEA 205.11 USD
ALA139194-100G ALA139194-1G ALA139194-25G ALA139194-5G
5-Amino-2-methylbenzenesulfonamide ≥98% (by HPLC)
5-Amino-2-methylbenzenesulphonamide
Formula: C₇H₁₀N₂O₂S
Melting Pt: 169 °C
MDL Number: MFCD06681071
CAS Number: 6973-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR