(3-Acetamidophenyl)boronic acid pinacol ester ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
N-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide, 3-Acetamidophenylboronic acid Pinacol ester, 3-Acetamidophenylboronic acid, pinacol estet, 2-(3-Acetamidophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acetanilide
A138890-1G
A138890-250MG
A138890-25G
A138890-5G
ALA138890-1GEA
48.36
USD
ALA138890-1G
ALA138890250MG
ALA138890-25G
ALA138890-5G
(3-Acetamidophenyl)boronic acid pinacol ester ≥97%
(3-Acetamidophenyl)boronic acid pinacol ester
Formula:
C₁₄H₂₀BNO₃ Melting Pt: 190 °C |
MDL Number:
MFCD03789261 CAS Number: 480424-93-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...