3-Acetylphenanthrene 90, technical

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 1-(phenanthren-3-yl)ethanone, 1-(phenanthren-3-yl)ethan-1-one, Ethanone, 1-(3-phenanthrenyl)-

A138450-1G A138450-250MG A138450-5G
ALA138450-1GEA 29.36 USD
ALA138450-1G ALA138450250MG ALA138450-5G
3-Acetylphenanthrene 90, technical
3-Acetylphenanthrene
Formula: C₁₆H₁₂O
MW: 220.27 g/mol
Melting Pt: 67-71°C
MDL Number: MFCD00001169
CAS Number: 2039-76-1
EINECS: 218-020-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2eaf3e73298f8160bb97ba196e8cd528704dfe6&vid=a2eaf3e73298f8160bb97ba196e8cd528704dfe6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR