1H-Pyrrolo[2,3-b]pyridine-3-carboxylicacid ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 7-Azaindole-3-carboxylic acid

A138304-100G A138304-1G A138304-25G
ALA138304-100GEA 3691.61 USD
ALA138304-100G ALA138304-1G ALA138304-25G
1H-Pyrrolo[2,3-b]pyridine-3-carboxylicacid ≥95%
1H-Pyrrolo[2,3-b]pyridine-3-carboxylicacid
Formula: C₈H₆N₂O₂
Melting Pt: 205-209°
MDL Number: MFCD07778360
CAS Number: 156270-06-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=906460722c94b9d5db17b3e3625aa30e19129f66&vid=906460722c94b9d5db17b3e3625aa30e19129f66&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR