5-Amino-1H-tetrazole monohydrate ≥99%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
5-Aminotetrazole monohydrate, 5-Amino-1H-tetrazole monohydrate, Tetrazol-5-amine, 1H-Tetrazol-5-ylamine monohydrate, 1H-Tetrazol-5-amine hydrate, 5-Amino-tetrazole-monohydrate
A138292-100G
A138292-25G
ALA138292-100GEA
49.31
USD
ALA138292-100G
ALA138292-25G
5-Amino-1H-tetrazole monohydrate ≥99%
5-Amino-1H-tetrazole monohydrate
Useful reagent in condensation reactions to form, for example, pyrimidines.
Formula:
CH₃N₅·1H₂O MW: 103.08 Melting Pt: 198-204°C Density: 1.51 |
MDL Number:
MFCD00149327 CAS Number: 15454-54-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...