Ethyl-2-aminoisonicotinate ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A138162-1G A138162-5G
ALA138162-1GEA 22.71 USD
ALA138162-1G ALA138162-5G
Ethyl-2-aminoisonicotinate ≥98%
Ethyl 2-aminoisonicotinate
Formula: C₈H₁₀N₂O₂
Melting Pt: 125 °C
MDL Number: MFCD03791260
CAS Number: 13362-30-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=047722326213d01f91ebb8fdf7061b89beb83e4a&vid=047722326213d01f91ebb8fdf7061b89beb83e4a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR