3-Amino-2-nitropyridine ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
2-Nitro-3-pyridylamine, 2-Nitropyridin-3-amine, 2-Nitropyridin-3-ylamine
A138155-1G
A138155-5G
ALA138155-1GEA
55.96
USD
ALA138155-1G
ALA138155-5G
3-Amino-2-nitropyridine ≥98%
3-Amino-2-nitropyridine
Formula:
C₅H₅N₃O₂ Melting Pt: 201 °C |
MDL Number:
MFCD00044102 CAS Number: 13269-19-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...