Auraptene ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: (E)-7-((3,7-Dimethylocta-2,6-dien-1-yl)oxy)-2H-chromen-2-one, 7-[[(2E)-3,7-Dimethyl-2,6-octadien-1-yl]oxy]-2H-chromen-2-one, 7-Geranyloxycoumarin

A137613-5MG A137613-100MG A137613-25MG
ALA137613-5MGEA 41.71 USD
ALA137613-5MG ALA137613100MG ALA137613-25MG
Auraptene ≥98% (by HPLC)
Auraptene

Auraptene (7-geranyloxycoumarin) is a prenyloxycoumarin, that is abundantly present in nature. It is produced by several plant species, such as Rutaceae and Umbeliferae (Apiaceae) families

Formula: C₁₉H₂₂O₃
MW: 298.4 g/mol
Melting Pt: 66°C
Storage Temperature: Freezer
MDL Number: MFCD00075948
CAS Number: 495-02-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b981df87a863ff71256bdf06f0992882b426362&vid=2b981df87a863ff71256bdf06f0992882b426362&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR