4-[[(4-Fluorophenyl)imino]methyl]phenol ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: ALPHA-(4-FLUOROPHENYLIMINO)-P-CRESOL, 4-[[(4-Fluorophenyl)imino]methyl]phenol

A137573-25G A137573-5G
ALA137573-25GEA 199.41 USD
ALA137573-25G ALA137573-5G
4-[[(4-Fluorophenyl)imino]methyl]phenol ≥98%
4-[[(4-Fluorophenyl)imino]methyl]phenol
Formula: C₁₃H₁₀FNO
Melting Pt: 181 °C
MDL Number: MFCD00029739
CAS Number: 3382-63-6
EINECS: 438-950-7, 636-494-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f88aca3bb96a804ac70e51ecc4a9de3c349dedbd&vid=f88aca3bb96a804ac70e51ecc4a9de3c349dedbd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR