2-Amino-2'-chloro-5-nitrobenzophenone ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Clonazepam Related Compound B, 2-Amino-5-nitro-2’-chlorobenzophenone, 2-Amino-2'-chloro-5-nitrobenzophenone, (2-Amino-5-nitrophenyl)(2-chlorophenyl)methanone

A137511-25G A137511-5G
ALA137511-25GEA 87.31 USD
ALA137511-25G ALA137511-5G
2-Amino-2'-chloro-5-nitrobenzophenone ≥98% (by GC)
2-Amino-2'-chloro-5-nitrobenzophenone
Formula: C₁₃H₉ClN₂O₃
Melting Pt: 121 °C
MDL Number: MFCD00792453
CAS Number: 2011-66-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ec92d37e8f65367688622bb02214ab3507a7a97&vid=6ec92d37e8f65367688622bb02214ab3507a7a97&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR