3-Acetamidocoumarin ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: N-(2-Oxo-2H-chromen-3-yl)acetamide

A137036-1G A137036-250MG
ALA137036-1GEA 85.41 USD
ALA137036-1G ALA137036250MG
3-Acetamidocoumarin ≥98%
3-Acetamidocoumarin
Formula: C₁₁H₉NO₃
Melting Pt: 208 °C
MDL Number: MFCD00075556
CAS Number: 779-30-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ba3db74eb8d97440394be906aba9f4f03bd963b&vid=3ba3db74eb8d97440394be906aba9f4f03bd963b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR