2-Aminoquinolin-8-ol ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A136838-1G A136838-5G
ALA136838-1GEA 98.71 USD
ALA136838-1G ALA136838-5G
2-Aminoquinolin-8-ol ≥98% (by GC)
2-Aminoquinolin-8-ol
Formula: C₉H₈N₂O
Melting Pt: 157-160 °C
MDL Number: MFCD00168931
CAS Number: 70125-16-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fd43afd3e07c886bded6e4f18a4b1005cb4f629&vid=6fd43afd3e07c886bded6e4f18a4b1005cb4f629&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR