2-Aminoquinoline ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

A136404-1G A136404-5G
ALA136404-1GEA 29.36 USD
ALA136404-1G ALA136404-5G
2-Aminoquinoline ≥97% (by GC)
2-Aminoquinoline
Formula: C₉H₈N₂
Melting Pt: 131 °C
MDL Number: MFCD00038079
CAS Number: 580-22-3
EINECS: 209-458-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff6debfe4cbb4d8541cfdcc2c85b5459483ed1a2&vid=ff6debfe4cbb4d8541cfdcc2c85b5459483ed1a2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR