ɑ-[2-(Methylamino)ethyl]benzyl alcohol 97

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Hydroxy-N-methyl-3-phenylpropylamine, 3-(Methylamino)-1-phenylpropanol, α-[2-(Methylamino)ethyl]benzyl Alcohol, 3-(Methylamino)-1-phenylpropan-1-ol

A136323-100G A136323-5G
ALA136323-100GEA 195.61 USD
ALA136323-100G ALA136323-5G
ɑ-[2-(Methylamino)ethyl]benzyl alcohol 97
alpha-[2-(Methylamino)ethyl]benzyl alcohol
Formula: C₁₀H₁₅NO
MW: 165.24 g/mol
Boiling Pt: 170°C
Melting Pt: 59-64 °C
Storage Temperature: Ambient
MDL Number: MFCD00674078
CAS Number: 42142-52-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35490463ed67420f9e7726c7a8c7efb42606c6d7&vid=35490463ed67420f9e7726c7a8c7efb42606c6d7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR