3-(Acetylthio)propionic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-(Acetylthio)propionicacid, S-Acetyl-3-mercaptopropionic acid, 3-(Acetylthio)propanoic acid

A136318-1G A136318-5G
ALA136318-1GEA 71.16 USD
ALA136318-1G ALA136318-5G
3-(Acetylthio)propionic acid ≥98%
3-(Acetylthio)propionic acid
Formula: C₅H₈O₃S
MW: 148.18 g/mol
Boiling Pt: 128 °C
Melting Pt: 54 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD06208418
CAS Number: 41345-70-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92b4078a8cace7d2f55f9c9498fbf2c43b8a131a&vid=92b4078a8cace7d2f55f9c9498fbf2c43b8a131a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR