2-Amino-5-hydroxymethylpyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Amino-5-pyridinemethanol, 6-Amino-3-pyridinemethanol, 2-Amino-5-(hydroxymethyl)pyridine, 2-Aminopyridine-5-methanol, (6-Aminopyridin-3-yl)methanol, (6-Amino-3-pyridyl)methanol

A136079-25G A136079-5G A136079-10G A136079-1G A136079-250MG
ALA136079-25GEA 1941.71 USD
ALA136079-25G ALA136079-5G ALA136079-10G ALA136079-1G ALA136079250MG
2-Amino-5-hydroxymethylpyridine ≥97%
2-Amino-5-hydroxymethylpyridine

Product Application:It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Formula: C₆H₈N₂O
MW: 124.14 g/mol
Melting Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD06659089
CAS Number: 113293-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b944828c3a2247e35569ced00d42560ab163ee71&vid=b944828c3a2247e35569ced00d42560ab163ee71&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR