5-Amino-2-naphthol ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A136056-1G A136056-25G A136056-5G
ALA136056-1GEA 36.96 USD
ALA136056-1G ALA136056-25G ALA136056-5G
5-Amino-2-naphthol ≥95%
5-Amino-2-naphthol

5-Amino-2-naphthol is a building block for proteomics research.A building block for proteomics research

Formula: C₁₀H₉NO
Melting Pt: 183 °C
MDL Number: MFCD00035729
CAS Number: 86-97-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f7de6270790012a05b23c8222c920bbb210987c&vid=4f7de6270790012a05b23c8222c920bbb210987c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR