Auraptene ≥95%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: (E)-7-((3,7-Dimethylocta-2,6-dien-1-yl)oxy)-2H-chromen-2-one, 7-[[(2E)-3,7-Dimethyl-2,6-octadien-1-yl]oxy]-2H-chromen-2-one, 7-Geranyloxycoumarin

A135842-25MG A135842-5MG
ALA135842-25MGEA 188.96 USD
ALA135842-25MG ALA135842-5MG
Auraptene ≥95%
Auraptene
Formula: C₁₉H₂₂O₃
MW: 298.4 g/mol
Melting Pt: 66°C
Storage Temperature: Ambient
MDL Number: MFCD00075948
CAS Number: 495-02-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e22e80933b4fb6b1cc4743fa6684d09d289742d&vid=3e22e80933b4fb6b1cc4743fa6684d09d289742d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR