9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate ≥99%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A135769-1G
A135769-5G
ALA135769-1GEA
36.01
USD
ALA135769-1G
ALA135769-5G
9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate ≥99%
9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate
Formula:
C₁₃H₁₄N₂·ClH·xH₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD00150067 CAS Number: 206658-92-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...